2-Nitro-4,6-bis(trifluoromethyl)aniline 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD257851-250MGEA 77.6 DKK
BLDPBD257851-250MG BLDPBD257851-1G
2-Nitro-4,6-bis(trifluoromethyl)aniline 97%
2-Nitro-4,6-bis(trifluoromethyl)aniline

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₈H₄F₆N₂O₂
Molekylvægt: 274,12 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD25962254
CAS-nummer: 320-21-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR