(4-(Bromomethyl)phenoxy)(tert-butyl)dimethylsilane 95+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD257734-1GEA 1230 DKK
BLDPBD257734-1G BLDPBD257734-5G BLDPBD257734-250MG
(4-(Bromomethyl)phenoxy)(tert-butyl)dimethylsilane 95+%
(4-(Bromomethyl)phenoxy)(tert-butyl)dimethylsilane

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₂₁BrOSi
Molekylvægt: 301,29 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD27920682
CAS-nummer: 120506-39-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR