3-Thiophencarbonitril 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD2575-5GEA 125 DKK
BLDPBD2575-5G BLDPBD2575-10G BLDPBD2575-25G BLDPBD2575-100G
3-Thiophencarbonitril 95%
3-Thiophencarbonitril

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₅H₃NS
Molekylvægt: 109,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00151852
CAS-nummer: 1641-09-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR