tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD257193-100MGEA 1050 DKK
BLDPBD257193-100MG BLDPBD257193-1G BLDPBD257193-5G BLDPBD257193-250MG
tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate 98%
tert-Butyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₂H₂₂N₂O₂
Molekylvægt: 226,32 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD17016673
CAS-nummer: 227940-72-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR