5-Aminoquinolin-2(1H)-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD256684-100MGEA 845 DKK
BLDPBD256684-100MG
5-Aminoquinolin-2(1H)-one 95%
5-Aminoquinolin-2(1H)-one

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₈N₂O
Molekylvægt: 160,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13705421
CAS-nummer: 61317-32-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR