4-(1H-Imidazol-2-yl)benzoic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD25668-1GEA 256 DKK
BLDPBD25668-1G BLDPBD25668-250MG BLDPBD25668-5G BLDPBD25668-10G BLDPBD25668-25G BLDPBD25668-100MG
4-(1H-Imidazol-2-yl)benzoic acid 97%
4-(1H-Imidazol-2-yl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₈N₂O₂
Molekylvægt: 188,183 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00456057
CAS-nummer: 108035-45-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR