4-Amino-2-chloropyridin-3-ol 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD256637-100MGEA 349 DKK
BLDPBD256637-100MG BLDPBD256637-1G BLDPBD256637-250MG
4-Amino-2-chloropyridin-3-ol 95%
4-Amino-2-chloropyridin-3-ol

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₅ClN₂O
Molekylvægt: 144,559 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16606120
CAS-nummer: 1227508-94-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR