Methyl 3-bromo-5-fluoroisonicotinate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD256519-5GEA 6050 DKK
BLDPBD256519-5G BLDPBD256519-250MG BLDPBD256519-100MG BLDPBD256519-1G
Methyl 3-bromo-5-fluoroisonicotinate 97%
Methyl 3-bromo-5-fluoroisonicotinate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₅BrFNO₂
Molekylvægt: 234,023 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14698118
CAS-nummer: 1214325-21-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR