6,7,8,9-Tetrahydro-5H-pyrrolo[2,3-b:5,4-c']dipyridine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD256476-50MGEA 752 DKK
BLDPBD256476-50MG
6,7,8,9-Tetrahydro-5H-pyrrolo[2,3-b:5,4-c']dipyridine hydrochloride 95%
6,7,8,9-Tetrahydro-5H-pyrrolo[2,3-b:5,4-c']dipyridine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₁₂ClN₃
Molekylvægt: 209,67 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26143076
CAS-nummer: 1354801-06-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR