3-Aminoadamantane-1-carboxylic acid hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD256096-1GEA 822 DKK
BLDPBD256096-1G BLDPBD256096-250MG BLDPBD256096-5G BLDPBD256096-100MG BLDPBD256096-25G
3-Aminoadamantane-1-carboxylic acid hydrochloride 95%
3-Aminoadamantane-1-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₈ClNO₂
Molekylvægt: 231,72 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01825827
CAS-nummer: 6240-01-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR