3-Methoxyazetidine hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD25596-5GEA 225 DKK
BLDPBD25596-5G BLDPBD25596-25G BLDPBD25596-1G BLDPBD25596-250MG BLDPBD25596-10G BLDPBD25596-100G
3-Methoxyazetidine hydrochloride 97%
3-Methoxyazetidine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C4H10ClNO
Molekylvægt: 123,58 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06804514
CAS-nummer: 148644-09-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR