6,7-Dimethyl-2,3-dihydro-1H-inden-1-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD255826-250MGEA 1700 DKK
BLDPBD255826-250MG BLDPBD255826-1G BLDPBD255826-100MG BLDPBD255826-5G
6,7-Dimethyl-2,3-dihydro-1H-inden-1-one 95%
6,7-Dimethyl-2,3-dihydro-1H-inden-1-one

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₂O
Molekylvægt: 160,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18647686
CAS-nummer: 16440-98-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR