(S)-1-(4-Isopropylphenyl)ethanamine hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD255647-50MGEA 690 DKK
BLDPBD255647-50MG BLDPBD255647-1G BLDPBD255647-100MG BLDPBD255647-250MG
(S)-1-(4-Isopropylphenyl)ethanamine hydrochloride 97%
(S)-1-(4-Isopropylphenyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₈ClN
Molekylvægt: 199,72 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16295045
CAS-nummer: 222737-88-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR