Methyl 5-bromo-2-(difluoromethoxy)benzoate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD255385-1GEA 714 DKK
BLDPBD255385-1G BLDPBD255385-5G BLDPBD255385-250MG
Methyl 5-bromo-2-(difluoromethoxy)benzoate 96%
Methyl 5-bromo-2-(difluoromethoxy)benzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇BrF₂O₃
Molekylvægt: 281,051 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11110847
CAS-nummer: 1131587-78-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR