3-Iodo-4-(piperidin-1-ylmethyl)benzoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD255305-100MGEA 473 DKK
BLDPBD255305-100MG BLDPBD255305-250MG BLDPBD255305-1G
3-Iodo-4-(piperidin-1-ylmethyl)benzoic acid 95%
3-Iodo-4-(piperidin-1-ylmethyl)benzoic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₃H₁₆INO₂
Molekylvægt: 345,17 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11110810
CAS-nummer: 1131614-61-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR