3-Boc-amino-4-bromopyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD25503-100MGEA 287 DKK
BLDPBD25503-100MG BLDPBD25503-250MG
3-Boc-amino-4-bromopyridine 95%
3-Boc-amino-4-bromopyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C10H13BrN2O2
Molekylvægt: 273,1300 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD05664029
CAS-nummer: 885275-14-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR