(R)-1-(3,4-Dimethylphenyl)propan-1-amine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254981-100MGEA 140 DKK
BLDPBD254981-100MG BLDPBD254981-1G BLDPBD254981-250MG
(R)-1-(3,4-Dimethylphenyl)propan-1-amine hydrochloride 95%
(R)-1-(3,4-Dimethylphenyl)propan-1-amine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₈ClN
Molekylvægt: 199,72 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12757583
CAS-nummer: 856563-05-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR