(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254955-5GEA 2670 DKK
BLDPBD254955-5G BLDPBD254955-1G BLDPBD254955-100MG BLDPBD254955-250MG
(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride 95%
(R)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₂₁ClN₂O₄
Molekylvægt: 280,75 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11101236
CAS-nummer: 1251903-83-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR