1,3-Dibromo-5-hexyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254717-100MGEA 489 DKK
BLDPBD254717-100MG
1,3-Dibromo-5-hexyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione 98%
1,3-Dibromo-5-hexyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₁₃Br₂NO₂S
Molekylvægt: 395,11 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26743584
CAS-nummer: 566939-56-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR