3,5-Dimethoxypyridine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD25464-100MGEA 85.3 DKK
BLDPBD25464-100MG BLDPBD25464-5G BLDPBD25464-25G BLDPBD25464-250MG BLDPBD25464-1G BLDPBD25464-10G
3,5-Dimethoxypyridine 97%
3,5-Dimethoxypyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₉NO₂
Molekylvægt: 139,152 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01646188
CAS-nummer: 18677-48-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR