5-Bromo-2-hydroxy-3-methylbenzonitrile 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254635-100MGEA 171 DKK
BLDPBD254635-100MG BLDPBD254635-1G BLDPBD254635-250MG BLDPBD254635-5G
5-Bromo-2-hydroxy-3-methylbenzonitrile 95%
5-Bromo-2-hydroxy-3-methylbenzonitrile

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₆BrNO
Molekylvægt: 212,043 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08457442
CAS-nummer: 913191-20-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR