4-Methoxy-1-naphthaleneboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254184-250MGEA 194 DKK
BLDPBD254184-250MG BLDPBD254184-25G BLDPBD254184-1G BLDPBD254184-5G
4-Methoxy-1-naphthaleneboronic acid 98%
4-Methoxy-1-naphthaleneboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₁H₁₁BO₃
Molekylvægt: 202,0142 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD03452757
CAS-nummer: 219834-95-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR