(5-Methylpyridin-2-yl)methanamine hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254165-250MGEA 528 DKK
BLDPBD254165-250MG BLDPBD254165-1G
(5-Methylpyridin-2-yl)methanamine hydrochloride 95%
(5-Methylpyridin-2-yl)methanamine hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₁₁ClN₂
Molekylvægt: 158,62 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD20731727
CAS-nummer: 161647-06-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR