Morpholine-2-carboxamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD254122-1GEA 1600 DKK
BLDPBD254122-1G BLDPBD254122-250MG BLDPBD254122-100MG BLDPBD254122-5G
Morpholine-2-carboxamide 95%
Morpholine-2-carboxamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₁₀N₂O₂
Molekylvægt: 130,14 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD15209643
CAS-nummer: 135072-13-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR