2-Ethoxy-5-fluoropyrimidin-4-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD253868-250MGEA 117 DKK
BLDPBD253868-250MG BLDPBD253868-1G
2-Ethoxy-5-fluoropyrimidin-4-amine 98%
2-Ethoxy-5-fluoropyrimidin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₈FN₃O
Molekylvægt: 157,146 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18973123
CAS-nummer: 56076-21-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR