4'-Bromo-2',6'-difluoroacetophenone 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD253863-100MGEA 69.8 DKK
BLDPBD253863-100MG BLDPBD253863-10G BLDPBD253863-25G BLDPBD253863-1G BLDPBD253863-250MG BLDPBD253863-5G
4'-Bromo-2',6'-difluoroacetophenone 97%
4'-Bromo-2',6'-difluoroacetophenone

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅BrF₂O
Molekylvægt: 235,026 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22570410
CAS-nummer: 746630-34-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR