7-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD253615-250MGEA 2280 DKK
BLDPBD253615-250MG BLDPBD253615-100MG
7-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
7-Bromo-4,4-dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₁H₁₄BrN·ClH
Molekylvægt: 276,601 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16618983
CAS-nummer: 1203684-66-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR