4-(2-Bromopyridin-4-yl)morpholine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252919-100MGEA 233 DKK
BLDPBD252919-100MG
4-(2-Bromopyridin-4-yl)morpholine 98%
4-(2-Bromopyridin-4-yl)morpholine

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₁₁BrN₂O
Molekylvægt: 243,1 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11869654
CAS-nummer: 1049023-41-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR