3-Ethyl-3-hydroxyazetidine hydrochloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252873-1GEA 2040 DKK
BLDPBD252873-1G BLDPBD252873-250MG BLDPBD252873-100MG
3-Ethyl-3-hydroxyazetidine hydrochloride 98%
3-Ethyl-3-hydroxyazetidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₂ClNO
Molekylvægt: 137,61 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11849053
CAS-nummer: 935668-00-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR