3-bromo-N-methylpyridin-2-amine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252836-1GEA 1420 DKK
BLDPBD252836-1G BLDPBD252836-5G BLDPBD252836-10G BLDPBD252836-250MG
3-bromo-N-methylpyridin-2-amine 98%
3-bromo-N-methylpyridin-2-amine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₇BrN₂
Molekylvægt: 187,037 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07374349
CAS-nummer: 214977-38-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR