2-Cyclopropoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252619-5GEA 15560 DKK
BLDPBD252619-5G BLDPBD252619-1G BLDPBD252619-250MG BLDPBD252619-100MG
2-Cyclopropoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 95%
2-Cyclopropoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₄H₂₀BNO₃
Molekylvægt: 261,12 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD13182227
CAS-nummer: 1346707-95-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR