2-Bromo-5-cyclopropylpyridine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252575-1GEA 3840 DKK
BLDPBD252575-1G BLDPBD252575-250MG BLDPBD252575-100MG
2-Bromo-5-cyclopropylpyridine 98%
2-Bromo-5-cyclopropylpyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₈BrN
Molekylvægt: 198,06 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11869551
CAS-nummer: 1142197-14-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR