(2-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252477-100MGEA 869 DKK
BLDPBD252477-100MG BLDPBD252477-5G BLDPBD252477-1G BLDPBD252477-250MG
(2-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%
(2-(Difluoromethoxy)pyridin-3-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₆BF₂NO₃
Molekylvægt: 188,92 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18261901
CAS-nummer: 1300750-50-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR