1-(Pyrimidin-2-yl)propan-1-one 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252295-100MGEA 388 DKK
BLDPBD252295-100MG BLDPBD252295-250MG BLDPBD252295-1G
1-(Pyrimidin-2-yl)propan-1-one 98%
1-(Pyrimidin-2-yl)propan-1-one

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₈N₂O
Molekylvægt: 136,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11847476
CAS-nummer: 54643-09-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR