N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD252003-100GEA 2030 DKK
BLDPBD252003-100G BLDPBD252003-25G BLDPBD252003-5G BLDPBD252003-10G
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine 98%
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine

Storage condition: Sealed in dry, 2-8°C

Formel: C₂₁H₂₅NO₅
Molekylvægt: 371,433 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00065701
CAS-nummer: 5545-54-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR