1,3,5-Tris(4-aminophenyl)benzene 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251965-250MGEA 210 DKK
BLDPBD251965-250MG BLDPBD251965-25G BLDPBD251965-1G BLDPBD251965-100MG BLDPBD251965-5G
1,3,5-Tris(4-aminophenyl)benzene 97%
1,3,5-Tris(4-aminophenyl)benzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₄H₂₁N₃
Molekylvægt: 351,45 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18207723
CAS-nummer: 118727-34-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR