3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251905-250MGEA 388 DKK
BLDPBD251905-250MG BLDPBD251905-5G BLDPBD251905-1G BLDPBD251905-10G
3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid 95%
3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₉NO₄
Molekylvægt: 229,27 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02093997
CAS-nummer: 855863-93-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR