Methyl 6-methylpyrazine-2-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251571-250MGEA 272 DKK
BLDPBD251571-250MG BLDPBD251571-100MG
Methyl 6-methylpyrazine-2-carboxylate 95%
Methyl 6-methylpyrazine-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₈N₂O₂
Molekylvægt: 152,151 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08705757
CAS-nummer: 41110-38-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR