Ethyl N-Boc-2,5-dihydropyrrole-3-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251426-100MGEA 179 DKK
BLDPBD251426-100MG BLDPBD251426-250MG
Ethyl N-Boc-2,5-dihydropyrrole-3-carboxylate 98%
Ethyl N-Boc-2,5-dihydropyrrole-3-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₂H₁₉NO₄
Molekylvægt: 241,28 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11973978
CAS-nummer: 146257-00-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR