1-Allyl-2-bromobenzene 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251351-1GEA 311 DKK
BLDPBD251351-1G BLDPBD251351-250MG BLDPBD251351-25G BLDPBD251351-5G
1-Allyl-2-bromobenzene 95%
1-Allyl-2-bromobenzene

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₉Br
Molekylvægt: 197,07 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01319538
CAS-nummer: 42918-20-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR