3,5,7-Trifluoroadamantane-1-carboxylic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251270-1GEA 791 DKK
BLDPBD251270-1G BLDPBD251270-250MG BLDPBD251270-5G BLDPBD251270-10G
3,5,7-Trifluoroadamantane-1-carboxylic acid 95%
3,5,7-Trifluoroadamantane-1-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₃F₃O₂
Molekylvægt: 234,215 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD17215150
CAS-nummer: 214557-89-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR