3-Hydroxy-2,2-dimethylpropyl isobutyrate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251170-1GEA 6210 DKK
BLDPBD251170-1G BLDPBD251170-250MG BLDPBD251170-100MG
3-Hydroxy-2,2-dimethylpropyl isobutyrate 95%
3-Hydroxy-2,2-dimethylpropyl isobutyrate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₈O₃
Molekylvægt: 174,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18974892
CAS-nummer: 5919-84-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR