5,5-Difluoropiperidine-3-carboxylic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251162-1GEA 5530 DKK
BLDPBD251162-1G BLDPBD251162-250MG BLDPBD251162-100MG
5,5-Difluoropiperidine-3-carboxylic acid 98%
5,5-Difluoropiperidine-3-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₆H₉F₂NO₂
Molekylvægt: 165,14 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18433628
CAS-nummer: 1255666-96-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR