2-Bromo-5-fluoroisonicotinonitrile 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251052-1GEA 2000 DKK
BLDPBD251052-1G BLDPBD251052-100MG BLDPBD251052-250MG
2-Bromo-5-fluoroisonicotinonitrile 95%
2-Bromo-5-fluoroisonicotinonitrile

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₂BrFN₂
Molekylvægt: 200,996 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18258644
CAS-nummer: 1256822-16-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR