2-Bromo-5-fluoro-3-methoxypyridine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD251051-5GEA 14340 DKK
BLDPBD251051-5G BLDPBD251051-100MG BLDPBD251051-1G BLDPBD251051-250MG
2-Bromo-5-fluoro-3-methoxypyridine 98%
2-Bromo-5-fluoro-3-methoxypyridine

Storage condition: Sealed in dry, 2-8°C

Formel: C₆H₅BrFNO
Molekylvægt: 206,01 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18257626
CAS-nummer: 1256806-73-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR