5-(2-Ethoxy-5-(piperazin-1-ylsulphonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250970-250MGEA 411 DKK
BLDPBD250970-250MG BLDPBD250970-5G BLDPBD250970-1G
5-(2-Ethoxy-5-(piperazin-1-ylsulphonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one 95%
5-(2-Ethoxy-5-(piperazin-1-ylsulphonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₂₁H₂₈N₆O₄S
Molekylvægt: 460,54 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00908398
CAS-nummer: 139755-82-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR