2-Chloro-5-ethoxypyrazine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250849-250MGEA 721 DKK
BLDPBD250849-250MG BLDPBD250849-100MG BLDPBD250849-1G
2-Chloro-5-ethoxypyrazine 95%
2-Chloro-5-ethoxypyrazine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₇ClN₂O
Molekylvægt: 158,58 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09607703
CAS-nummer: 136309-11-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR