Methyl 6-methylpyrimidine-4-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250819-100MGEA 256 DKK
BLDPBD250819-100MG BLDPBD250819-250MG
Methyl 6-methylpyrimidine-4-carboxylate 98%
Methyl 6-methylpyrimidine-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₈N₂O₂
Molekylvægt: 152,151 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13174781
CAS-nummer: 73955-53-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR