Benzyl (trans-4-aminocyclohexyl)carbamate hydrochloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250415-1GEA 551 DKK
BLDPBD250415-1G BLDPBD250415-250MG BLDPBD250415-25G BLDPBD250415-5G
Benzyl (trans-4-aminocyclohexyl)carbamate hydrochloride 98%
Benzyl (trans-4-aminocyclohexyl)carbamate hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₄H₂₁ClN₂O₂
Molekylvægt: 284,78 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11007681
CAS-nummer: 1217664-37-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR