4-Borono-3-fluorothioanisole 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD250263-250MGEA 512 DKK
BLDPBD250263-250MG BLDPBD250263-100MG
4-Borono-3-fluorothioanisole 98%
4-Borono-3-fluorothioanisole

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₈BFO₂S
Molekylvægt: 186,0116 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09027247
CAS-nummer: 957060-84-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR